Learn More
Thermo Scientific Chemicals Neomycin sulfate, 50 mg/ml in distilled water, sterile-filtered
$167.45 - $255.67
Chemical Identifiers
CAS | 1405-10-3 |
---|---|
Molecular Formula | C23H48N6O17S |
Molecular Weight (g/mol) | 712.72 |
MDL Number | MFCD29905465 |
InChI Key | OIXVKQDWLFHVGR-DJUBFIBWNA-N |
PubChem CID | 124080932 |
IUPAC Name | 5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid |
SMILES | OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67011AD
|
Thermo Scientific Chemicals
J67011AD |
50 mL |
Each for $167.45
|
|
|||||
AAJ67011AE
|
Thermo Scientific Chemicals
J67011AE |
100 mL |
Each for $255.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1405-10-3 | |
712.72 | |
OIXVKQDWLFHVGR-DJUBFIBWNA-N | |
5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid |
C23H48N6O17S | |
MFCD29905465 | |
124080932 | |
OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O |
Specifications
Neomycin Sulfate | |
1405-10-3 | |
MFCD29905465 | |
OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O | |
712.72 | |
50 mL |
Liquid | |
C23H48N6O17S | |
OIXVKQDWLFHVGR-DJUBFIBWNA-N | |
5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid | |
124080932 | |
Neomycin sulfate, 50mg/mL in distilled water, sterile-filtered |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H361d
EINECSNumber : 215-773-1
TSCA : Yes
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only