Please login to your online account to display your discounted pricing

Neodymium standard solution, for AAS, 1mg/mL Nd in 2% HNO3, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Neodymium standard solution
Name Note 1 mg/ml Nd in 2% HNO3
CAS 7440-00-8
CAS 7697-37-2
CAS Min % 1.9
CAS Max % 2.1
CAS 7732-18-5
CAS Min % 96.9
CAS Max % 97.1
Purity Grade For AAS
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC196141000

  SDS
thermo fisher scientific
196141000
Plastic bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.24
Physical Form Solution
Purity Grade For AAS
Quantity 100mL
Color Clear
MDL Number MFCD00011130
Name Note 1 mg/ml Nd in 2% HNO3
Chemical Name or Material Neodymium standard solution
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 1.9
CAS 7697-37-2
CAS 7732-18-5
CAS Min % 96.9
CAS Max % 97.1
CAS Max % 2.1
Packaging Plastic bottle
Concentration 995 to 1005 μg/mL of Nd in 2% HNO3 (20°C)
Health Hazard 1 GHS Signal Word: Warning
CAS 7440-00-8
Molecular Formula Nd
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

for atomic adsorption spectroscopy

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.