missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Neodymium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLND22Y
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Nd | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Neodymium Standard | |
Triple-leached LDPE bottle | |
0.1,2.0,97.9 | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011130 | |
[Nd] | |
144.24 | |
CHEBI:33372 | |
Matrix Concentration Cat. No. 2-5% HNO31000μg/mLPLND2-2Y, -2X 10,000 PLND2-3Y, -3X |
7440-00-8,7697-37-2,7732-18-5 | |
125 mL | |
0.1,2.0,97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Nd | |
QEFYFXOXNSNQGX-UHFFFAOYSA-N | |
neodymium | |
23934 | |
Assurance | |
2% HNO3 |
Chemical Identifiers
7440-00-8 | |
144.24 | |
QEFYFXOXNSNQGX-UHFFFAOYSA-N | |
CHEBI:33372 | |
[Nd] |
Nd | |
MFCD00011130 | |
23934 | |
neodymium |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause an allergic skin reaction. Suspected of causing cancer.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only