missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium rod, 6.35mm (0.25 in.) dia., 99.5% (metals basis excluding Ta)

Manufacturer:  Alfa Aesar™ 44051HF

 View more versions of this product

Catalog No. AA44051HF



Description

Description

  • Packaged in mylar under argon
Specifications

Specifications

Neodymium rod
10 mm
7440-00-8
Nd
atom
[Nd]
Odorless
Mylar under Argon
CHEBI:33372
99.5%
≈2.2g/mm
≈6.35mm (0.25 in.) dia.
(metals basis excluding Ta)
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodymium
144.242
23934
≈0.22g/mm,Cast Rod,diameter is approximate

Chemical Identifiers

7440-00-8
144.242
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
neodymium
Nd
MFCD00011130
atom
CHEBI:33372
[Nd]
Safety and Handling

Safety and Handling

EINECSNumber : 231-109-3

RTECSNumber : QO8575000

TSCA : Yes

storageNote1 :

  • Packaged in mylar under argon

Recommended Storage : Ambient temperatures

Documents