missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals
$218.12 - $1390.73
Chemical Identifiers
CAS | 7440-00-8 |
---|---|
Molecular Formula | Nd |
Molecular Weight (g/mol) | 144.24 |
MDL Number | MFCD00011130 |
InChI Key | QEFYFXOXNSNQGX-UHFFFAOYSA-N |
Synonym | neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n |
PubChem CID | 23934 |
ChEBI | CHEBI:33372 |
IUPAC Name | neodymium |
SMILES | [Nd] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA40294HF
|
Thermo Scientific Chemicals
040294HF |
10 mm |
Each for $218.12
|
|
|||||
AA40294DH
|
Thermo Scientific Chemicals
040294DH |
25 mm |
Each for $371.52
|
|
|||||
AA40294DJ
|
Thermo Scientific Chemicals
040294DJ |
50 mm |
Each for $681.77
|
|
|||||
AA40294DM
|
Thermo Scientific Chemicals
040294DM |
100 mm |
Each for $1,390.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7440-00-8 | |
144.24 | |
QEFYFXOXNSNQGX-UHFFFAOYSA-N | |
23934 | |
neodymium |
Nd | |
MFCD00011130 | |
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n | |
CHEBI:33372 | |
[Nd] |
Specifications
7440-00-8 | |
MFCD00011130 | |
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n | |
[Nd] | |
144.24 | |
CHEBI:33372 | |
Odorless | |
≈0.89g/mm | |
Neodymium rod, 12.7mm (0.5 in.) dia. |
Nd | |
10 mm | |
QEFYFXOXNSNQGX-UHFFFAOYSA-N | |
neodymium | |
23934 | |
99.5% | |
Mylar under Argon | |
Cast Rod | |
(metals basis excluding Ta) |
Safety and Handling
EINECSNumber : 231-109-3
RTECSNumber : QO8575000
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only