missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 040294HF

 View more versions of this product

Catalog No. AA40294HF



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Neodymium rod
7440-00-8
Nd
10 mm
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodymium
23934
99.5%
Mylar under Argon
Cast Rod
12.7mm (0.5 in.) dia.
(metals basis excluding Ta)
MFCD00011130
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
[Nd]
144.24
CHEBI:33372
Odorless
≈0.89g/mm

Chemical Identifiers

7440-00-8
144.24
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
neodymium
Nd
MFCD00011130
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
CHEBI:33372
[Nd]
Safety and Handling

Safety and Handling

EINECSNumber : 231-109-3

RTECSNumber : QO8575000

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only