missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium powder, -40 mesh, 99.8% (REO)

Manufacturer:  Alfa Aesar™ 4418106

 View more versions of this product

Catalog No. AA4418106



Description

Description

  • Ampouled under argon
Specifications

Specifications

Neodymium powder
7440-00-8
Nd
UN3089
atom
[Nd]
Odorless
Ampule
CHEBI:33372
99.9%
Gray to Silver
5 g
(REO)
MFCD00011130
−40 Mesh
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodymium
144.242
23934
Powder
Air and moisture sensitive

Chemical Identifiers

7440-00-8
144.242
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
neodymium
Nd
MFCD00011130
atom
CHEBI:33372
[Nd]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P235+P410-P240-P241-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q

H228-H315-H319-X

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

EINECSNumber : 231-109-3

RTECSNumber : QO8575000

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Argon

Documents