missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium powder, -40 mesh, 99.8% (REO)

Manufacturer:  Alfa Aesar™ 4418114

 View more versions of this product

Catalog No. AA4418114



Description

Description

  • Ampouled under argon
Specifications

Specifications

Neodymium powder
Air & Moisture Sensitive
25g
Odorless
(REO)
MFCD00011130
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
[Nd]
144.242
CHEBI:33372
99.9%
−40 Mesh
Gray to Silver
7440-00-8
Ampule
Nd
UN3089
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodymium
23934
Powder

Chemical Identifiers

7440-00-8
144.242
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
neodymium
Nd
MFCD00011130
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
CHEBI:33372
[Nd]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378b-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Special powder for metal fires.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

RTECSNumber : QO8575000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents