missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium powder, -200 mesh, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 4591406

 View more versions of this product

Catalog No. AA4591406



Description

Description

  • Packaged under Argon
Specifications

Specifications

Neodymium powder
Air & Moisture Sensitive
5g
Odorless
Nd
UN1383
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodymium
23934
Powder
−200 Mesh
Gray to Silver
7440-00-8
(REO)
MFCD00011130
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
[Nd]
144.242
CHEBI:33372
99.9%

Chemical Identifiers

7440-00-8
144.242
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
neodymium
Nd
MFCD00011130
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
CHEBI:33372
[Nd]
Safety and Handling

Safety and Handling

GHS H Statement
H250-H260-H252

GHS P Statement
P210-P222-P231+P232-P280-P235+P410-P233-P422a-P420-P407-P501a

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC METALS, N.O.S.

RTECSNumber : QO8575000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents