missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neodymium(III) Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T191925G

 View more versions of this product

Catalog No. T191925G



Specifications

Specifications

Neodymium(III) Trifluoromethanesulfonate
25g
C3F9NdO9S3
1759
WYRSPTDNOIZOGA-UHFFFAOYSA-K
neodymium(3+);trifluoromethanesulfonate
2733937
Crystal-Powder at 20°C
Purple
34622-08-7
MFCD00192343
neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3]
591.432
591.43
≥97.0% (T)

Chemical Identifiers

34622-08-7
591.432
WYRSPTDNOIZOGA-UHFFFAOYSA-K
2733937
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Nd+3]
C3F9NdO9S3
MFCD00192343
neodymium iii trifluoromethanesulfonate, neodymium iii triflate, neodymium trifluoromethanesulfonate, trifluoromethanesulfonic acid neodymium iii salt, nd otf 3, neodymium triflate, acmc-209i8y, neodymium 3+ ion tritriflate, neodymium perfluoromethanesulfonate, neodymium 3+ ; trifluoromethanesulfonate
neodymium(3+);trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents