missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium(III) titanate

$361.73 - $361.73

Chemical Identifiers

CAS 12058-94-5
Molecular Formula Nd2Ti2O7
Formula Weight 496.21
MDL Number MFCD00192342
Synonym Neodymium titanium oxide
Chemical Name or Material Neodymium(III) titanate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1736614 SDS Alfa Aesar™
A1736614
25g Each for $361.73
AAA1736606 SDS Alfa Aesar™
A1736606
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

12058-94-5
496.21
Neodymium titanium oxide
Nd2Ti2O7
MFCD00192342
Neodymium(III) titanate

Specifications

12058-94-5
496.21
Neodymium titanium oxide
25g
Nd2Ti2O7
MFCD00192342
Odorless
Neodymium(III) titanate