Learn More
Neodymium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04025314
Description
It is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Neodymium(III) sulfate octahydrate | |
Rare Earth Oxides Content | |
13477-91-3 | |
25 g | |
Nd2(SO4)3·8H2O | |
Soluble in water | |
Odorless | |
2.85 g/cm3 at 20°C |
1176°C | |
Pink | |
Crystals | |
99.9% | |
MFCD00149803 | |
720.78 (576.66 Anhydrous) | |
Pink crystals |
Chemical Identifiers
13477-91-3 | |
Nd2(SO4)3·8H2O | |
MFCD00149803 |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 233-262-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only