missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium(III) oxide, REacton™, 99.997% (REO)

Manufacturer:  Alfa Aesar™ 1383714

 View more versions of this product

Catalog No. AA1383714



Specifications

Specifications

Neodymium(III) oxide
(REO)
14,6450
Powder
Reacton™
2272°C
1313-97-9
MFCD00011134
336.48
99.997%
7.24g/mL
25g

Chemical Identifiers

1313-97-9
MFCD00011134
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents