missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium(III) nitrate hexahydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1291218

 View more versions of this product

Catalog No. AA1291218



Specifications

Specifications

Neodymium(III) nitrate hexahydrate
(REO)
MFCD00149799
14,6450
VQVDTKCSDUNYBO-UHFFFAOYSA-N
neodymium(3+);trinitrate;hexahydrate
204494
Crystalline
Odorless
~40°C
50g
16454-60-7
H12N3NdO15
UN1477
neodymium nitrate hexahydrate, neodymium trinitrate hexahydrate, neodymium nitrate monohydrate, neodymium iii nitrate, hexahydrate 1:3:6, nitric acid, neodymium 3+ salt, hexahydrate, ksc491s0r, neodymiumnitratehexahydrate, neodymium nitrate 6-hydrate, neodymium 3+ trinitrate hexahydrate, neodymium iii nitrate hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3]
438.344
438.34 (330.25 Anhydrous)
99.9%
Hygroscopic
3.0 to 4.0

Chemical Identifiers

16454-60-7
438.344
VQVDTKCSDUNYBO-UHFFFAOYSA-N
204494
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Nd+3]
H12N3NdO15
MFCD00149799
neodymium nitrate hexahydrate, neodymium trinitrate hexahydrate, neodymium nitrate monohydrate, neodymium iii nitrate, hexahydrate 1:3:6, nitric acid, neodymium 3+ salt, hexahydrate, ksc491s0r, neodymiumnitratehexahydrate, neodymium nitrate 6-hydrate, neodymium 3+ trinitrate hexahydrate, neodymium iii nitrate hexahydrate
neodymium(3+);trinitrate;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : OJ6125000

TSCA : TSCA

Documents