Learn More
Neodymium(III) iodide, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04715706
Description
Neodymium(III) iodide is used in glass coloring, permanent magnets, capacitors, infrared filters, resonators, catalysis (diene polymerization), laser materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Neodymium(III) iodide | |
Black | |
Powder | |
(metals basis) | |
Soluble in water. | |
99.9% | |
Ampule |
Ultra dry | |
13813-24-6 | |
5 g | |
MFCD00016248 | |
524.95 | |
Odorless | |
Hygroscopic; Light sensitive |
Chemical Identifiers
13813-24-6 | |
MFCD00016248 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 237-467-7
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only