Please login to your online account to display your discounted pricing

Neodymium(III) fluoride 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Neodymium(III) fluoride
CAS 13709-42-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (ex fluor)
Assay Percent Range 98%
Molecular Formula F3Nd
Linear Formula NdF3
Formula Weight 201.23
MDL Number MFCD00011135
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383140500

  SDS
thermo fisher scientific
AC383140500
Plastic bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (ex fluor)
Formula Weight 201.23
Physical Form Powder
Quantity 50g
Color Light gray
CAS Max % 100.0
CAS 13709-42-7
Linear Formula NdF3
Molecular Formula F3Nd
Assay Percent Range 98%
CAS Min % 97.5
MDL Number MFCD00011135
Chemical Name or Material Neodymium(III) fluoride
Packaging Plastic bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with acids liberates very toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.