missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium(III) acetate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 4458718

 View more versions of this product

Catalog No. AA4458718



Specifications

Specifications

Neodymium (III) acetate hydrate
(REO)
MFCD00013040
321.38 (Anhydrous)
99.9%
334869-71-5
Nd(OOCCH3)3·xH2O
Soluble in water,moderately soluble in strong mineral acids.
Crystalline
50g

Chemical Identifiers

334869-71-5
Nd(OOCCH3)3·xH2O
MFCD00013040
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents