Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium Gallium Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 110 Orientation, Alfa Aesar™

$303.24 - $1,333.01

Chemical Identifiers

Chemical Name or Material Neodymium Gallium Oxide Substrate
Name Note Polished One Side
Molecular Formula NdGaO3
MDL Number MFCD00210638
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38504KT

 
alfa aesar™
38504KT
1 ea. Each for $303.24

AA38504KU

 
alfa aesar™
38504KU
5 ea. Each for $1,333.01
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Neodymium Gallium Oxide Substrate
Name Note Polished One Side
Molecular Formula NdGaO3
MDL Number MFCD00210638