missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium foil, 1.0mm (0.04 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 10248FF

 View more versions of this product

Catalog No. AA10248FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Neodymium foil
25 x 25mm
7440-00-8
Mylar under Argon
Nd
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
[Nd]
144.242
CHEBI:33372
99.9%
≈4.4g/25x25mm
1.0mm (0.04 in.) thick
Odorless
(REO)
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodymium
23934
Foil

Chemical Identifiers

7440-00-8
144.242
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
neodymium
Nd
MFCD00011130
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
CHEBI:33372
[Nd]
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents