Please login to your online account to display your discounted pricing

Alfa Aesar™ Neodymium boride, 99.5%

Chemical Identifiers

Chemical Name or Material Neodymium boride
CAS 12008-23-0
Formula Weight 209.10g/mol
MDL Number MFCD00151359
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3949406

 
alfa aesar™
39494-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Mesh Size −325 Mesh
Odor Odorless
Quantity 5g
Quantity 5g
Chemical Name or Material Neodymium boride
CAS 12008-23-0
Assay 99.5%
Formula Weight 209.10g/mol
MDL Number MFCD00151359
TSCA TSCA