Please login to your online account to display your discounted pricing

Neocuproine Reagent, 0.1% (w/v) in Methanol, APHA for Copper, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Neocuproine Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5240-16

 
ricca chemical company
RI524016
Plastic 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5240-4

 
ricca chemical company
RI52404
Plastic 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Chemical Name or Material Neocuproine Reagent
Packaging Plastic
Methods APHA for Copper
Concentration or Composition (by Analyte or Components) 0.1% (w/v) in Methanol