Please login to your online account to display your discounted pricing

Neocuproine Reagent, 0.1% in Methanol, for Copper, LabChem™

$67.69 - $67.69

Chemical Identifiers

Chemical Name or Material Neocuproine Reagent
CAS 67-56-1
CAS Max % 99.9
CAS 654054-57-6
CAS Max % 0.1
Name Note 0.1% in Methanol, for Copper
Linear Formula C14H14N2O
Formula Weight 226.27
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC176307

 
lab chem
LC176307
125mL Each for $67.69
Description & Specifications

Specifications

Formula Weight 226.27
Chemical Name or Material Neocuproine Reagent
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Boiling Point 65°C
Color Clear, colorless to slight yellow
Melting Point −98°C
Quantity 125mL
Flash Point 9.7°C
Name Note 0.1% in Methanol, for Copper
Physical Form Liquid
Linear Formula C14H14N2O
Density 0.79g/mL
CAS 67-56-1
UN Number UN1993
Health Hazard 1 Danger
CAS Max % 99.9
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, kidneys, liver, optic nerve).
Identification Passes Test
CAS 654054-57-6
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 0.1
Solubility Soluble in water
Specific Gravity 0.79g/mL
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, kidneys, liver, optic nerve).