missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neo/GUIDE Microscope Slide Sets

Manufacturer:  Fisher Scientific AZ704210

 View more versions of this product

Catalog No. S67723



Includes: all slides from the following sets described above: Mitosis and Meiosis, Five Kingdoms, Survey of Algae, Plant Anatomy, Survey of Animals, and Human Body.

Description

Description

Sets come complete with Neo/GUIDE reference booklet featuring detailed background information and structure identification key. Available as single sets or classroom packs of multiple sets.

Common Things (Set of 5 slides)
•    Feather (three types)
•    Housefly head, leg, and wing
•    Textile fibers
•    Hair (three types)
•    Letter “e”
Cells (Set of 5)
•    Bacteria (Prokaryotic cell)
•    Cheek (Eukaryotic cell)
•    Cyanobacteria (Photosynthetic bacteria)
•    Mixed protozoa
•    Lilac leaf (plant cell)
Microlife (Set of 5)
•    Bacteria
•    Bread mold
•    Yeast
•    Planktonic forms
•    Mixed protozoa
Bacteria (One slide)
•    Contains smears of cocci, bacilli and spirilla on one slide
Protists (Set of 5)
•    Paramecium
•    Amoeba
•    Spirogyra
•    Mixed protists
•    Euglena
Plants (Set of 5)
•    Onion mitosis
•    Plant root
•    Plant stem
•    Plant leaf
•    Plant flower bud
Invertebrates (Set of 5)
•    Daphnia
•    Planaria
•    Hydra
•    Plankton
•    Rotifers
Disease-causing Organisms (Set of 5)
•    Trichinella (Trichinosis)
•    Plasmodium (Malaria)
•    Pneumocystis (AIDS-opportunistic infection)
•    Mycrobacterium (Tuberculosis)
•    Entamoeba (Amoebic dysentery)
Fungi (set of 5)
•    Aspergillus
•    Molds (three types)
•    Mushroom
•    Bread mold
•    Sporigia and zygotes
•    Budding yeast
Exploring Life, Set I (41 slides)
Includes all slides from Common Things, Cells, Microlife, Bacteria, Protists, Plants, Invertebrates, Disease-causing Organisms, and Fungi Sets, described above.
Survey of Algae (Set of 10 slides)
•    Euglena, wm
•    Volvox, wm
•    Chlamydomonas, wm
•    Spirogyra—conjugation, wm
•    Oedogonium, wm
•    Fucus, cs
•    Porphyra, wm
Five Kingdoms (set of 20)
•    Monera—Mixed bacteria, Anabaena, Oscillatoria
•    Protista—Vorticella, Paramecium, Amoeba, Volvox
•    Fungi—Penicillium, Rhizopus Stolonifer
•    Plantae—Moss leaf, Pinus leaf, Fern (Prothallium), Helianthus stem
•    Animalia—Hydra, Planaria, Bed bug, Earthworm, Sea urchin, Fish scales, Frog skin
Mitosis and Meiosis (Set of 5)
•    Onion root tip mitosis
•    Whitefish blastula mitosis
•    Meiosis I in plant cells
•    Meiosis II in plant cells
•    Meiosis in animal cells (Grasshopper Testis)
Human Body Systems (Set of 20)
•    Tissues—Squamous Epithelium, Adipose tissue, Striated muscle
•    Skeletal—Compact bone, Hyaline cartilage
•    Nervous—Nerve cells, Spinal cord, Cerebellum
•    Circulatory—Artery and Vein, Blood smear (Giemsa stain)
•    Organs—Liver, Stomach, Small intestine, Lung, Spleen, Colon, Thyroid Gland, Pancreas, Human skin, Kidney
Comparing Plant Anatomy (Set of 5)
•    Monocot and dicot root
•    Monocot and dicot stem
•    Monocot and dicot leaf
•    Monocot and dicot flower bud
•    Deciduous and evergreen leaf
Survey of Animals (Set of 20)
•    Grantia
•    Hydra
•    Planaria
•    Liver fluke
•    Taenia pisiformis
•    Ascaris
•    Earthworm
•    Daphnia
•    Spider (Head and mouthparts)
•    Mites
•    Coackroach (Head and mouthparts)
•    Locust (Mouthparts)
•    Locust (Thorax)
•    Locust (Abdomen)
•    Drosophila (Wild type)
•    Culex (Pupa)
•    Aphid
•    Sea urchin (Ova)
•    Fish scales
•    Frog skin
Exploring Life, Set II (80 slides)

Specifications

Specifications

A microscopic investigation of disease-causing organisms
Exploring Disease-Causing Organisms Slide Set
Class set: 5 sets of 5 slides
Prepared
SDS
Documents