Invitrogen™ NEK9 Recombinant Human Protein DFS Item
NEK9 belongs to the NIMA family of kinases.
Manufacturer:  Invitrogen™ LSPV4653
 View more versions of this product
Catalog No. PV4653
Description
Description
The NIMA family of kinases are involved in DNA replication and genotoxic stress response.
Specifications
Specifications
NP_149107.2
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100
92.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic, Recombinant
NEK9, NERC, NERCC, NEK8
Kinase Assay
Baculovirus
aa 347-732
Yes
Cell Viability, Proliferation & Function|DNA Replication & Repair|Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Cell Analysis|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
91754
10μg
RUO - research use only
Serine/Threonine Kinases
NIMA (never in mitosis gene a)- related kinase 9
Kinases & Inhibitors
NEK Kinase Family
Human
GST Tag
Documents