Invitrogen™ NEK4 Recombinant Human Protein DFS Item
NEK4 is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4331
 View more versions of this product
Catalog No. PV4331
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
NEK4 is involved in signal transduction and cellular communication. It is proposed to act exclusively upon threonine residues and is present in most primary carcinomas.
Specifications
Specifications
NP_003148
Frozen
122kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
NEK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
NEK4, NRK2
Baculovirus
Human
GST Tag
Documents