missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ NEK11 (FLJ23495) Recombinant Human Protein
NEK11 (FLJ23495) belongs to the NIMA family of kinases.
Supplier: Invitrogen™ PV4648
Description
NEK11 (FLJ23495) is involved in DNA replication and genotoxic stress response.
Specifications
NP_079076 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
101.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
NEK Kinase Family | |
Recombinant | |
Baculovirus |
Cell Analysis, DNA Replication and Repair, Nuclear Receptor Assay | |
79858 | |
1mg | |
RUO - research use only | |
NEK11 (FLJ23495) | |
Human | |
GST Tag | |
Frozen |