NEK1 Recombinant Human Protein DFS Item
NEK1 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4202
 View more versions of this product
Catalog No. PV4202
Description
Description
NEK1 is a serine⁄threonine protein kinase, related to the NIMA cell cycle regulator, and is highly expressed in meiotic germ cells. The kinase may function early in the DNA damage response pathway. NEK1 may also participate in polycystic kidney disease and neural cell development.
Specifications
Specifications
NP_036356
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
85.3kDa
85% by SDS-PAGE
–80°C
1 Each
NEK1
NEK1 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
NEK
NEK1 Recombinant Human Protein
Human
0.34mg/mL
4750, 84634
NEK1 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
NEK1 Recombinant Human Protein
Kinase assay
Baculovirus
Amino acids 1-505
590nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL
GST Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.