missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nefazodone hydrochloride, 98+%

$139.70 - $595.26

Chemical Identifiers

CAS 82752-99-6
Molecular Formula C25H33Cl2N5O2
Molecular Weight (g/mol) 506.472
MDL Number MFCD00935760
InChI Key DYCKFEBIOUQECE-UHFFFAOYSA-N
Synonym nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, nefazodone hydrochloride, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
PubChem CID 54911
ChEBI CHEBI:7495
IUPAC Name 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-5-ethyl-4-(2-phenoxyethyl)-1,2,4-triazol-3-one;hydrochloride
SMILES CCC1=NN(C(=O)N1CCOC2=CC=CC=C2)CCCN3CCN(CC3)C4=CC(=CC=C4)Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6279303 View Documents Alfa Aesar™
J6279303
1g Each for $139.70
 
AAJ6279306 View Documents Alfa Aesar™
J6279306
5g Each for $595.26
 
Description

Description

  • An antidepressant acts by modifying serotonin transmission
  • Mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor
Specifications

Chemical Identifiers

82752-99-6
506.472
DYCKFEBIOUQECE-UHFFFAOYSA-N
54911
2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-5-ethyl-4-(2-phenoxyethyl)-1,2,4-triazol-3-one;hydrochloride
C25H33Cl2N5O2
MFCD00935760
nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, nefazodone hydrochloride, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
CHEBI:7495
CCC1=NN(C(=O)N1CCOC2=CC=CC=C2)CCCN3CCN(CC3)C4=CC(=CC=C4)Cl.Cl

Specifications

White
1g
C25H33Cl2N5O2
14,6438
DYCKFEBIOUQECE-UHFFFAOYSA-N
2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-5-ethyl-4-(2-phenoxyethyl)-1,2,4-triazol-3-one;hydrochloride
54911
506.47
≥98%
186°C to 187°C
82752-99-6
MFCD00935760
nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, nefazodone hydrochloride, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
CCC1=NN(C(=O)N1CCOC2=CC=CC=C2)CCCN3CCN(CC3)C4=CC(=CC=C4)Cl.Cl
506.472
CHEBI:7495
Powder
Nefazodone hydrochloride