Please login to your online account to display your discounted pricing

NECA, Tocris

Adenosine receptor agonist

$75.29 - $75.29

Chemical Identifiers

Chemical Name or Material NECA
Assay Percent Range >99%
Molecular Formula C12H16N6O4
Formula Weight 308.3
Synonym 1-(6-Amino-9H-purin-9-yl)-1-deoxy-N-ethyl-β-D-ribofuranuronamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

16-911-0

 
r & d systems
16911
10mg Each for $75.29
Description & Specifications

Specifications

Formula Weight 308.3
Quantity 10mg
Chemical Name or Material NECA
Synonym 1-(6-Amino-9H-purin-9-yl)-1-deoxy-N-ethyl-β-D-ribofuranuronamide
Assay Percent Range >99%
Molecular Formula C12H16N6O4
CAS 35920-39-9
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C