Please login to your online account to display your discounted pricing

Nebivolol hydrochloride, Tocris™

Highly selective β1-adrenoceptor antagonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material Nebivolol hydrochloride
Assay Percent Range >98%
Molecular Formula C22H25F2NO4·HCl
Formula Weight 441.9
Synonym (±)-[2R™(1S™5S™(S™))]-α, α'-[Iminobis(methylene)bis(6-fluoro-3, 4-dihydro-2H-1-benzopyran-2-methanol)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

351710

 
r & d systems
RDS351710
10mg Each for $214.12

351750

 
r & d systems
RDS351750
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 441.9
Quantity 10mg
Chemical Name or Material Nebivolol hydrochloride
Assay Percent Range >98%
Molecular Formula C22H25F2NO4·HCl
CAS 152520-56-4
Synonym (±)-[2R™(1S™5S™(S™))]-α, α'-[Iminobis(methylene)bis(6-fluoro-3, 4-dihydro-2H-1-benzopyran-2-methanol)
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature