missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nebivolol Hydrochloride, TCI America™

$206.25 - $713.48

Chemical Identifiers

CAS 152520-56-4
Molecular Formula C22H26ClF2NO4
Molecular Weight (g/mol) 445.924
MDL Number MFCD07357273
InChI Key JWEXHQAEWHKGCW-PZFCLNQZSA-N
PubChem CID 131674192
IUPAC Name (1S)-2,2-dideuterio-2-[[(2S)-1,1-dideuterio-2-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-hydroxyethyl]amino]-1-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)ethanol;hydrochloride
SMILES C1CC2=C(C=CC(=C2)F)OC1C(CNCC(C3CCC4=C(O3)C=CC(=C4)F)O)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0954100MG View Documents TCI America
N0954100MG
100mg Each for $713.48
 
N095420MG View Documents TCI America
N095420MG
20mg Each for $206.25
 
Specifications

Chemical Identifiers

152520-56-4
445.924
JWEXHQAEWHKGCW-PZFCLNQZSA-N
(1S)-2,2-dideuterio-2-[[(2S)-1,1-dideuterio-2-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-hydroxyethyl]amino]-1-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)ethanol;hydrochloride
C22H26ClF2NO4
MFCD07357273
131674192
C1CC2=C(C=CC(=C2)F)OC1C(CNCC(C3CCC4=C(O3)C=CC(=C4)F)O)O.Cl

Specifications

White
100mg
C22H26ClF2NO4
JWEXHQAEWHKGCW-PZFCLNQZSA-N
(1S)-2,2-dideuterio-2-[[(2S)-1,1-dideuterio-2-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-hydroxyethyl]amino]-1-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)ethanol;hydrochloride
131674192
Crystal-Powder at 20°C
Nebivolol Hydrochloride
226°C
152520-56-4
MFCD07357273
C1CC2=C(C=CC(=C2)F)OC1C(CNCC(C3CCC4=C(O3)C=CC(=C4)F)O)O.Cl
445.924
441.90
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No