Nebivolol Hydrochloride 98.0+%, TCI America™
Supplier:  TCI America N0954100MG
 View more versions of this product
Catalog No. N0954100MG
Description
Description
Specifications
Specifications
Nebivolol Hydrochloride
100mg
C22H26ClF2NO4
Crystalline Powder at 20°C
≥98.0% (HPLC)
152520-56-4
226°C
441.90
White
Chemical Identifiers
152520-56-4
C22H26ClF2NO4