Please login to your online account to display your discounted pricing

Alfa Aesar™ Ndelta-Boc-Nalpha-Fmoc-D-ornithine, 95%

Chemical Identifiers

Chemical Name or Material Ndelta-Boc-Nalpha-Fmoc-D-ornithine
CAS 118476-89-4
Formula Weight 454.52
MDL Number MFCD00077065
Synonym Fmoc-D-Orn(Boc)-OH; (R)-5-(Boc-amino)-2-(Fmoc-amino)pentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6293003

  SDS
alfa aesar™
H6293003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6293006

  SDS
alfa aesar™
H6293006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 454.52
Synonym Fmoc-D-Orn(Boc)-OH; (R)-5-(Boc-amino)-2-(Fmoc-amino)pentanoic acid
Color Yellow
Physical Form Powder
MDL Number MFCD00077065
Chemical Name or Material Ndelta-Boc-Nalpha-Fmoc-D-ornithine
Assay 95%
CAS 118476-89-4
Quantity 1g