missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ndelta-Boc-Nalpha-Fmoc-D-ornithine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6293006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ndelta-Boc-Nalpha-Fmoc-D-ornithine | |
Yellow | |
MFCD00077065 | |
JOOIZTMAHNLNHE-OAQYLSRUSA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid | |
13996154 | |
95% | |
Powder |
118476-89-4 | |
C25H30N2O6 | |
fmoc-d-orn boc-oh, n-fmoc-n'-boc-d-ornithine, 2r-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, fmoc-n'-boc-d-ornithine, ambotzfaa1492, fmoc-nd-boc-d-ornithine, n-alpha-fmoc-nd-t-boc-d-ornithine, r-n5-tert.-butoxy-carbonyl-n2-9-fluorenylmethoxycarbonyl-ornithine, 2r-5-tert-butoxycarbonylamino-2-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid | |
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
454.523 | |
454.52 | |
5 g |
Chemical Identifiers
118476-89-4 | |
454.523 | |
JOOIZTMAHNLNHE-OAQYLSRUSA-N | |
13996154 | |
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
C25H30N2O6 | |
MFCD00077065 | |
fmoc-d-orn boc-oh, n-fmoc-n'-boc-d-ornithine, 2r-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, fmoc-n'-boc-d-ornithine, ambotzfaa1492, fmoc-nd-boc-d-ornithine, n-alpha-fmoc-nd-t-boc-d-ornithine, r-n5-tert.-butoxy-carbonyl-n2-9-fluorenylmethoxycarbonyl-ornithine, 2r-5-tert-butoxycarbonylamino-2-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only