Please login to your online account to display your discounted pricing

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 98%

Chemical Identifiers

Chemical Name or Material Ndelta-Benzyloxycarbonyl-Nalpha-Fmoc-L-ornithine
CAS 138775-07-2
Formula Weight 488.54
MDL Number MFCD00190893
Synonym Fmoc-Orn(Z)-OH; (S)-5-(Benzyloxycarbonylamino)-2-(Fmoc-amino)pentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6659806

  SDS
alfa aesar™
H6659806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6659814

  SDS
alfa aesar™
H6659814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6659822

  SDS
alfa aesar™
H6659822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 488.54
CAS 138775-07-2
Physical Form Powder
Assay 98%
Synonym Fmoc-Orn(Z)-OH; (S)-5-(Benzyloxycarbonylamino)-2-(Fmoc-amino)pentanoic acid
Storage Note 1 -30° to -10°C
Chemical Name or Material Ndelta-Benzyloxycarbonyl-Nalpha-Fmoc-L-ornithine
MDL Number MFCD00190893
Color Off-white to white
Quantity 5g
Safety and Handling

-30° to -10°C