missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-Nalpha-Boc-L-ornithine, 98%

$28.68 - $274.50

Chemical Identifiers

CAS 2480-93-5
Molecular Formula C18H26N2O6
Molecular Weight (g/mol) 366.414
MDL Number MFCD00038259
InChI Key QYYCZJUFHDLLOJ-AWEZNQCLSA-N
Synonym boc-orn z-oh, s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-orn cbz-oh, nalpha-boc-ndelta-cbz-l-ornithine, nalpha-tert-butoxycarbonyl-ndelta-carbobenzoxy-l-ornithine, 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-phenylmethoxycarbonylamino pentanoic acid, 2s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-tert-butoxycarbonyl-n'-benzyloxycarbonyl-l-ornithine, n-alpha-boc-n-delta-z-l-ornithine, bocnh-orn z-oh
PubChem CID 7269338
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6226803 View Documents Alfa Aesar™
H6226803
1g Each for $28.68
 
AAH6226806 View Documents Alfa Aesar™
H6226806
5g Each for $79.88
 
AAH6226814 View Documents Alfa Aesar™
H6226814
25g Each for $274.50
 
Specifications

Chemical Identifiers

2480-93-5
366.414
QYYCZJUFHDLLOJ-AWEZNQCLSA-N
7269338
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O
C18H26N2O6
MFCD00038259
boc-orn z-oh, s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-orn cbz-oh, nalpha-boc-ndelta-cbz-l-ornithine, nalpha-tert-butoxycarbonyl-ndelta-carbobenzoxy-l-ornithine, 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-phenylmethoxycarbonylamino pentanoic acid, 2s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-tert-butoxycarbonyl-n'-benzyloxycarbonyl-l-ornithine, n-alpha-boc-n-delta-z-l-ornithine, bocnh-orn z-oh
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

2480-93-5
MFCD00038259
boc-orn z-oh, s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-orn cbz-oh, nalpha-boc-ndelta-cbz-l-ornithine, nalpha-tert-butoxycarbonyl-ndelta-carbobenzoxy-l-ornithine, 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-phenylmethoxycarbonylamino pentanoic acid, 2s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-tert-butoxycarbonyl-n'-benzyloxycarbonyl-l-ornithine, n-alpha-boc-n-delta-z-l-ornithine, bocnh-orn z-oh
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O
366.414
366.41
1g
C18H26N2O6
2783761
QYYCZJUFHDLLOJ-AWEZNQCLSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
7269338
98%
Ndelta-Benzyloxycarbonyl-Nalpha-Boc-L-ornithine