missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine, 98%

$24.36 - $336.18

Chemical Identifiers

CAS 16937-92-1
Molecular Formula C18H26N2O6
Molecular Weight (g/mol) 366.414
MDL Number MFCD00069970
InChI Key QYYCZJUFHDLLOJ-CQSZACIVSA-N
Synonym boc-d-orn z-oh, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, n, a-z-n
PubChem CID 7272030
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63925MD View Documents Alfa Aesar™
H63925MD
250mg Each for $24.36
 
AAH6392503 View Documents Alfa Aesar™
H6392503
1g Each for $81.73
 
AAH6392506 View Documents Alfa Aesar™
H6392506
5g Each for $336.18
 
Specifications

Chemical Identifiers

16937-92-1
366.414
QYYCZJUFHDLLOJ-CQSZACIVSA-N
7272030
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O
C18H26N2O6
MFCD00069970
boc-d-orn z-oh, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, n, a-z-n
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

16937-92-1
MFCD00069970
boc-d-orn z-oh, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, n, a-z-n
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O
366.414
366.41
98%
Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine
C18H26N2O6
2400891
QYYCZJUFHDLLOJ-CQSZACIVSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
7272030
Solid
250mg
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C