missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6392506
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine | |
C18H26N2O6 | |
2400891 | |
QYYCZJUFHDLLOJ-UHFFFAOYNA-N | |
5-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid | |
7272030 | |
98% | |
Solid |
16937-92-1 | |
MFCD00069970 | |
boc-d-orn z-oh, nalpha-boc-ndelta-cbz-r-ornithine, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, a-boc-n, ksc918o2r, a-z-n, a-boc-n, a-z-d-ornithine, a-z-n, a-boc-d-ornithine | |
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O | |
366.41 | |
366.41 | |
5 g |
Chemical Identifiers
16937-92-1 | |
366.41 | |
QYYCZJUFHDLLOJ-UHFFFAOYNA-N | |
7272030 |
C18H26N2O6 | |
MFCD00069970 | |
boc-d-orn z-oh, nalpha-boc-ndelta-cbz-r-ornithine, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, a-boc-n, ksc918o2r, a-z-n, a-boc-n, a-z-d-ornithine, a-z-n, a-boc-d-ornithine | |
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold