missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine, 98%

Manufacturer:  Alfa Aesar™ H63925MD

 View more versions of this product

Catalog No. AAH63925MD



Specifications

Specifications

Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine
C18H26N2O6
2400891
QYYCZJUFHDLLOJ-CQSZACIVSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
7272030
Solid
250mg
16937-92-1
MFCD00069970
boc-d-orn z-oh, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, n, a-z-n
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O
366.414
366.41
98%

Chemical Identifiers

16937-92-1
366.414
QYYCZJUFHDLLOJ-CQSZACIVSA-N
7272030
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O
C18H26N2O6
MFCD00069970
boc-d-orn z-oh, a-boc-d-ornithine, nalpha-boc-ndelta-cbz-r-ornithine, boc-d-orn z-oh tlc, 2r-5-benzyloxycarbonylamino-2-tert-butoxycarbonylamino pentanoic acid, r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-boc-n'-cbz-d-ornithine, 2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, ksc918o2r, n, a-boc-n, a-z-d-ornithine, n, a-z-n
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents