missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride, 98%

$29.62 - $457.77

Chemical Identifiers

CAS 5874-75-9
Molecular Formula C14H21ClN2O4
Molecular Weight (g/mol) 316.782
MDL Number MFCD00077027
InChI Key ZUJMKNWYEVTCNT-YDALLXLXSA-N
Synonym c14h20n2o4.hcl, h-orn cbz-ome inverted exclamation mark currencyhcl, h-orn z-ome hcl, h-orn z-ome hydrochloride, h-orn z-ome-hcl, h-orn z-ome.hcl, l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1, methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride, methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride, n-delta-z-l-ornithine methyl ester hydrochloride
PubChem CID 13967650
IUPAC Name methyl (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoate;hydrochloride
SMILES COC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6564703 View Documents Alfa Aesar™
H6564703
1g Each for $29.62
 
AAH6564706 View Documents Alfa Aesar™
H6564706
5g Each for $100.36
 
AAH6564714 View Documents Alfa Aesar™
H6564714
25g Each for $457.77
 
Specifications

Chemical Identifiers

5874-75-9
316.782
ZUJMKNWYEVTCNT-YDALLXLXSA-N
13967650
COC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)N.Cl
C14H21ClN2O4
MFCD00077027
c14h20n2o4.hcl, h-orn cbz-ome inverted exclamation mark currencyhcl, h-orn z-ome hcl, h-orn z-ome hydrochloride, h-orn z-ome-hcl, h-orn z-ome.hcl, l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1, methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride, methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride, n-delta-z-l-ornithine methyl ester hydrochloride
methyl (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoate;hydrochloride

Specifications

5874-75-9
MFCD00077027
c14h20n2o4.hcl, h-orn cbz-ome inverted exclamation mark currencyhcl, h-orn z-ome hcl, h-orn z-ome hydrochloride, h-orn z-ome-hcl, h-orn z-ome.hcl, l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1, methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride, methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride, n-delta-z-l-ornithine methyl ester hydrochloride
COC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)N.Cl
316.782
316.78
98%
1g
C14H21ClN2O4
4341483
ZUJMKNWYEVTCNT-YDALLXLXSA-N
methyl (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoate;hydrochloride
13967650
Solid
138°C to 141°C
Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold