missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H6564706

 View more versions of this product

Catalog No. AAH6564706



Specifications

Specifications

Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride
C14H21ClN2O4
4341483
ZUJMKNWYEVTCNT-YDALLXLXSA-N
methyl (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoate;hydrochloride
13967650
Solid
138°C to 141°C
5874-75-9
MFCD00077027
h-orn z-ome.hcl, h-orn z-ome hcl, h-orn z-ome-hcl, h-orn z-ome hydrochloride, c14h20n2o4.hcl, n-delta-z-l-ornithine methyl ester hydrochloride, h-orn cbz-ome inverted exclamation mark currencyhcl, methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride, l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1, methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride
COC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)N.Cl
316.782
316.78
98%
5g

Chemical Identifiers

5874-75-9
316.782
ZUJMKNWYEVTCNT-YDALLXLXSA-N
13967650
COC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)N.Cl
C14H21ClN2O4
MFCD00077027
h-orn z-ome.hcl, h-orn z-ome hcl, h-orn z-ome-hcl, h-orn z-ome hydrochloride, c14h20n2o4.hcl, n-delta-z-l-ornithine methyl ester hydrochloride, h-orn cbz-ome inverted exclamation mark currencyhcl, methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride, l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1, methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride
methyl (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoate;hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents