Please login to your online account to display your discounted pricing

Alfa Aesar™ Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 95%

Chemical Identifiers

Chemical Name or Material Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine
CAS 147290-11-7
Formula Weight 438.47
MDL Number MFCD00798637
Synonym Fmoc-Orn(Alloc)-OH; (S)-5-(Allyloxycarbonylamino)-2-(Fmoc-amino)pentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6333103

 
alfa aesar™
H6333103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6333106

 
alfa aesar™
H6333106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6333114

 
alfa aesar™
H6333114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 438.47
Chemical Name or Material Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine
Physical Form Solid
Assay 95%
MDL Number MFCD00798637
CAS 147290-11-7
Synonym Fmoc-Orn(Alloc)-OH; (S)-5-(Allyloxycarbonylamino)-2-(Fmoc-amino)pentanoic acid
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C