Please login to your online account to display your discounted pricing

NCX 4040, Tocris™

NO-donating aspirin; anti-inflammatory and antitumor

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material NCX 4040
Assay Percent Range >98%
Molecular Formula C16H13NO7
Formula Weight 331.28
Synonym 2-(Acetyloxy)benzoic acid 4-[(nitrooxy)methyl]phenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

453110

 
r & d systems
RDS453110
10mg Each for $205.70

453150

 
r & d systems
RDS453150
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 331.28
Quantity 10mg
Assay Percent Range >98%
CAS 287118-97-2
Synonym 2-(Acetyloxy)benzoic acid 4-[(nitrooxy)methyl]phenyl ester
Recommended Storage Store at -20°C
Chemical Name or Material NCX 4040
Molecular Formula C16H13NO7
Safety and Handling

Store at -20°C