Please login to your online account to display your discounted pricing

NCS-382, Tocris

Antagonist of γ-hydroxybutyric acid

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material NCS-382
Assay Percent Range >99%
Molecular Formula C13H14O3
Formula Weight 218.25
Synonym 6,7,8,9-Tetrahydro-5-hydroxy-5H-benzocyclohept-6-ylideneacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

078010

 
r & d systems
RDS078010
10mg Each for $146.74

078050

 
r & d systems
RDS078050
50mg Each for $618.87
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 218.25
CAS 520505-01-5
Assay Percent Range >99%
Synonym 6,7,8,9-Tetrahydro-5-hydroxy-5H-benzocyclohept-6-ylideneacetic acid
Molecular Formula C13H14O3
Chemical Name or Material NCS-382
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C