Please login to your online account to display your discounted pricing

NCH 51, Tocris™

Histone deacetylase inhibitor

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material NCH 51
Assay Percent Range >98%
Molecular Formula C20H26N2O2S2
Formula Weight 390.56
Synonym 2-Methylpropanethioic acid S-[7-oxo-7-[(4-phenyl-2-thiazolyl)amino]heptyl] ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

374710

 
r & d systems
RDS374710
10mg Each for $214.12

374750

 
r & d systems
RDS374750
50mg Each for $895.76
Description & Specifications

Specifications

Chemical Name or Material NCH 51
Quantity 10mg
CAS 848354-66-5
Recommended Storage Store at +4°C
Assay Percent Range >98%
Molecular Formula C20H26N2O2S2
Formula Weight 390.56
Synonym 2-Methylpropanethioic acid S-[7-oxo-7-[(4-phenyl-2-thiazolyl)amino]heptyl] ester
Safety and Handling

Store at +4°C