Please login to your online account to display your discounted pricing

NBQX, Tocris


Potent AMPA antagonist; More selective than CNQX

Quantity 10mg

Manufacturer: r & d systems  037310

 View more versions of this product

Catalog No. 037310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 336.28
CAS 118876-58-7
Molecular Formula C12H8N4O6S
Chemical Name or Material NBQX
Assay Percent Range >98%
Synonym 2,3-Dioxo-6-nitro-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulfonamide
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material NBQX
Assay Percent Range >98%
Molecular Formula C12H8N4O6S
Formula Weight 336.28
Synonym 2,3-Dioxo-6-nitro-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulfonamide
Accessories

Store at Room Temperature