Fisherbrand™ Natural Polypropylene Jars with White Polypropylene Unlined Cap
Ideal for storing and collecting specimens and lab supplies. Fisherbrand™ Natural Polypropylene Jars with White Polypropylene Unlined Cap are translucent and durable.
$48.03 - $112.87
Specifications
Shape Straight Sided Jar
Mouth Wide
Material Polypropylene
Color Natural
Closure Color White
View More Specs
Includes: White Polypropylene unlined caps
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
02-912-024A Fisherbrand™
ALP263491
89-400 24/Cs. 16 oz. 480mL Case of 24 Each for $73.90
02-912-025A Fisherbrand™
ALP263492
43-400 72/Cs. 1 oz. 30mL Case of 72 Each for $85.77
02-912-027A Fisherbrand™
ALP263495
120-400 24/Cs. 32 oz. 960mL Case of 24 Each for $112.87
02-912-026A Fisherbrand™
ALP263494
53-400 48/Cs. 2 oz. 60mL Case of 48 Each for $48.03
02-912-028A Fisherbrand™
ALP263497
58-400 36/Cs. 4 oz. 120mL Case of 36 Each for $49.23
02-912-029A Fisherbrand™
ALP263499
89-400 36/Cs. 8 oz. 240mL Case of 36 Each for $90.47
Specifications
Description
  • Features wide neck and straight sides to facilitate easy recovery of contents
  • Polypropylene material offers greater tolerance to high temperatures than HDPE and better contact clarity
Description & Specifications
Specifications
Straight Sided Jar
Polypropylene
White
Polypropylene
Wide
Natural
Screw
Unlined