Please login to your online account to display your discounted pricing
Fisherbrand™ Natural Polypropylene Jars without Cap
Ideal for storing and collecting specimens and lab supplies. Fisherbrand™ Natural Polypropylene Jars without Cap are translucent and durable.
Manufacturer: fisherbrand™  ALP263496
 View more versions of this product
Catalog No. 02-912-261A
  • / Case of 432 Each
Description & Specifications
Specifications
Capacity (Metric) 120mL
Capacity (English) 4 oz.
Product Type Straight Sided Jar
Color Natural
Material Polypropylene
Mouth Wide
Neck Finish 58-400
Quantity 432/Cs.
Shape Straight Sided Jar
  • Features wide neck and straight sides to facilitate easy recovery of contents
  • Polypropylene material offers tolerance to high temperatures than HDPE and better contact clarity