Please login to your online account to display your discounted pricing
Fisherbrand™ Natural Polypropylene Jars without Cap
Ideal for storing and collecting specimens and lab supplies. Fisherbrand™ Natural Polypropylene Jars without Cap are translucent and durable.
$225.13 - $297.27
Specifications
Color Natural
Material Polypropylene
Mouth Wide
Shape Straight Sided Jar
View More Specs
Products
Catalog Number Mfr. No. Quantity Capacity (English) Capacity (Metric) Neck Finish Price Quantity    
02-912-260A
fisherbrand™
ALP263493
546/Cs. 2 oz. 60mL 53-400 Case of 546 Each for $231.47
02-912-258A
fisherbrand™
ALP263498
168/Cs. 8 oz. 240mL 89-400 Case of 168 Each for $225.13
02-912-261A
fisherbrand™
ALP263496
432/Cs. 4 oz. 120mL 58-400 Case of 432 Each for $297.27
Description & Specifications
Specifications
Color Natural
Material Polypropylene
Mouth Wide
Shape Straight Sided Jar
  • Features wide neck and straight sides to facilitate easy recovery of contents
  • Polypropylene material offers tolerance to high temperatures than HDPE and better contact clarity
Caps not included