Fisherbrand™ Natural LDPE Boston Round Bottles Black with Phenolic PolyCone Lined Cap
Ideal solution for liquid sample handling in a squeezable bottle. Fisherbrand™ Natural LDPE Boston Round Bottles Black with Phenolic PolyCone Lined Cap are unbreakable and more resistant to stress cracking than HDPE bottles.
$65.30 - $157.60
Specifications
Liner Material PolyCone
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Natural
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
02-911-909A Fisherbrand™
ALP263425
28-400 24/Cs. 16 oz. 480mL Case of 24 Each for $107.57
02-911-966A Fisherbrand™
ALP263437
24-410 48/Cs. 4 oz. 120mL Case of 48 Each for $100.97
02-911-967A Fisherbrand™
ALP263440
24-410 48/Cs. 8 oz. 240mL Case of 48 Each for $157.60
02-912-015A Fisherbrand™
ALP263427
18-410 48/Cs. 1 oz. 30mL Case of 48 Each for $65.30
Specifications
Description
  • Bottles retain their impact strength at very low temperatures
  • Caps form an tight seal and offer a good chemical barrier
  • Liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
Description & Specifications
Specifications
PolyCone
Screw
Natural
Narrow
Phenolic
Black
LDPE
Round